Assay Details
Gene Symbol: LOC686129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008767400.1 | XP_008765622.1 | 1 | 912 | 69 |
Gene Symbol: Bmpr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030849.1 | NP_110476.1 | 7 - 8 | 901 | 69 | |
GenBank mRNA | D17667.1 | - | 6 - 7 | 676 | 69 | |
D38082.1 | - | 6 - 7 | 758 | 69 | ||
S75359.1 | - | 7 - 8 | 901 | 69 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
mesoderm formation
somitogenesis
Mullerian duct regression
regulation of endothelial cell proliferation
positive regulation of mesenchymal cell proliferation
heart morphogenesis
endocardial cushion formation
transcription by RNA polymerase II
protein phosphorylation
immune response
pattern specification process
ectoderm development
nervous system development
endoderm development
heart development
biological_process
dorsal/ventral axis specification
anterior/posterior pattern specification
dorsal/ventral pattern formation
positive regulation of pathway-restricted SMAD protein phosphorylation
neural crest cell development
stem cell population maintenance
pituitary gland development
neural plate mediolateral regionalization
cell differentiation
lung development
positive regulation of bone mineralization
BMP signaling pathway
hindlimb morphogenesis
odontogenesis of dentin-containing tooth
embryonic digit morphogenesis
regulation of endothelial cell differentiation
positive regulation of osteoblast differentiation
positive regulation of transcription, DNA-templated
paraxial mesoderm development
paraxial mesoderm structural organization
lateral mesoderm development
regulation of lateral mesodermal cell fate specification
mesendoderm development
embryonic organ development
developmental growth
embryonic morphogenesis
positive regulation of epithelial cell proliferation
negative regulation of neurogenesis
cartilage development
roof of mouth development
positive regulation of SMAD protein signal transduction
neural plate pattern specification
heart formation
regulation of cellular senescence
DNA-binding transcription factor activity, RNA polymerase II-specific
molecular_function
protein serine/threonine kinase activity
transforming growth factor beta-activated receptor activity
ATP binding
protein homodimerization activity
SMAD binding
metal ion binding
in utero embryonic development
mesoderm formation
somitogenesis
Mullerian duct regression
regulation of endothelial cell proliferation
positive regulation of mesenchymal cell proliferation
heart morphogenesis
endocardial cushion formation
transcription by RNA polymerase II
protein phosphorylation
immune response
pattern specification process
ectoderm development
nervous system development
endoderm development
heart development
biological_process
dorsal/ventral axis specification
anterior/posterior pattern specification
dorsal/ventral pattern formation
positive regulation of pathway-restricted SMAD protein phosphorylation
neural crest cell development
stem cell population maintenance
pituitary gland development
neural plate mediolateral regionalization
cell differentiation
lung development
positive regulation of bone mineralization
BMP signaling pathway
hindlimb morphogenesis
odontogenesis of dentin-containing tooth
embryonic digit morphogenesis
regulation of endothelial cell differentiation
positive regulation of osteoblast differentiation
positive regulation of transcription, DNA-templated
paraxial mesoderm development
paraxial mesoderm structural organization
lateral mesoderm development
regulation of lateral mesodermal cell fate specification
mesendoderm development
embryonic organ development
developmental growth
embryonic morphogenesis
positive regulation of epithelial cell proliferation
negative regulation of neurogenesis
cartilage development
roof of mouth development
positive regulation of SMAD protein signal transduction
neural plate pattern specification
heart formation
regulation of cellular senescence
DNA-binding transcription factor activity, RNA polymerase II-specific
molecular_function
protein serine/threonine kinase activity
transforming growth factor beta-activated receptor activity
ATP binding
protein homodimerization activity
SMAD binding
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P357470 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342102 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138935 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197756 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175718 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876901 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521278 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542491 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504465 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634009 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355751 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942883 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165799 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674206 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760293 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209526 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388628 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58886 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344586 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756369 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303358 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701713 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915070 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78072 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108077 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124976 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75389 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163989 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178939 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P357470 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P357470 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P357470 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P455656 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P357470 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700252202 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700251973 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 32062 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 8580 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 55791 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700252202 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700251973 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10790487 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10790484 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10790482 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10790489 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10790483 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10790488 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10790486 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10790485 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10790481 | - | - |
Rat Genome 230 2.0 Array | 1390398_PM_at | - | - |
Rat Genome 230 2.0 Array | 1390398_at | - | - |
Rat Genome 230 2.0 Array | 1368899_PM_at | - | - |
Rat Genome 230 2.0 Array | 1368899_at | - | - |
Affymetrix HT Rat Focus Array | 1368899_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368899_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1600009504.rn.1 | TC1600001237.rn.1 | RTA TC |
Rat Expression Set 230 | 1368899_PM_at | - | - |
Rat Expression Set 230 | 1368899_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10790481 | - | - |
Rat Genome U34 Set | S75359_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21489207 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373335 | - | - |
RatRef-12 Expression BeadChip | ILMN_59030 | - | - |
