Assay Details
Gene Symbol: Parg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031339.1 | NP_112629.1 | 10 - 11 | 2190 | 150 | |
XM_006252669.1 | XP_006252731.1 | 9 - 10 | 1860 | 150 | ||
GenBank mRNA | AB019366.1 | - | 10 - 11 | 2190 | 150 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
