Assay Details
Gene Symbol: Parg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031339.1 | NP_112629.1 | 15 - 16 | 2660 | 115 | |
XM_006252669.1 | XP_006252731.1 | 14 - 15 | 2330 | 115 | ||
GenBank mRNA | AB019366.1 | - | 15 - 16 | 2660 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
