Assay Details
Gene Symbol: Glt8d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007683.1 | NP_001007684.1 | 1 - 2 | 141 | 101 | |
GenBank mRNA | BC079066.1 | - | 1 - 2 | 141 | 101 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC079066.1, NM_001007683.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
