Assay Details
Gene Symbol: Il17rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107290.1 | NP_001100760.1 | 2 - 3 | 182 | 74 | |
XM_006252603.2 | XP_006252665.1 | 2 - 3 | 168 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107290.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
