Assay Details
Gene Symbol: Il17rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107290.1 | NP_001100760.1 | 7 - 8 | 775 | 107 | |
XM_006252603.2 | XP_006252665.1 | 7 - 8 | 757 | 107 | ||
GenBank mRNA | BC168886.1 | - | 3 - 4 | 320 | 7369142 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
