Assay Details
Gene Symbol: Il17rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107290.1 | NP_001100760.1 | 5 - 6 | 583 | 109 | |
XM_006252603.2 | XP_006252665.1 | 5 - 6 | 565 | 109 | ||
GenBank mRNA | BC168886.1 | - | 1 - 2 | 128 | 7369142 | 109 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC168886.1,NM_001107290.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
