Assay Details
Gene Symbol: Mettl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007623.1 | NP_001007624.1 | 1 - 2 | 105 | 110 | |
GenBank mRNA | BC079309.1 | - | 1 - 2 | 105 | 110 | |
FQ211422.1 | - | 1 - 2 | 113 | 110 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC079309.1, NM_001007623.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
