Assay Details
Gene Symbol: Mettl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007623.1 | NP_001007624.1 | 4 - 5 | 765 | 85 | |
GenBank mRNA | BC079309.1 | - | 4 - 5 | 765 | 85 | |
FQ211422.1 | - | 4 - 5 | 774 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
