Assay Details
Gene Symbol: Gmds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039606.1 | NP_001034695.1 | 9 - 10 | 1005 | 99 | |
GenBank mRNA | BC104708.1 | - | 9 - 10 | 1005 | 7317156 | 99 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
