Assay Details
Gene Symbol: Elovl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109118.1 | NP_001102588.1 | 2 - 3 | 194 | 122 | |
XM_006253779.2 | XP_006253841.1 | 2 - 3 | 140 | 122 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001109118.1, XM_006253779.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
