Assay Details
Gene Symbol: Elovl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109118.1 | NP_001102588.1 | 3 - 4 | 375 | 79 | |
XM_006253779.2 | XP_006253841.1 | 3 - 4 | 321 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
