Assay Details
Gene Symbol: Nol7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253849.2 | XP_006253911.1 | 4 - 5 | 543 | 78 | |
GenBank mRNA | FQ210031.1 | - | 4 - 5 | 415 | 78 | |
FQ227425.1 | - | 4 - 5 | 411 | 78 | ||
FQ229485.1 | - | 4 - 5 | 425 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
