Assay Details
Gene Symbol: Nol7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253849.2 | XP_006253911.1 | 6 - 7 | 759 | 95 | |
GenBank mRNA | FQ210031.1 | - | 6 - 7 | 631 | 95 | |
FQ227425.1 | - | 6 - 7 | 627 | 95 | ||
FQ229485.1 | - | 6 - 7 | 641 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
