Assay Details
Gene Symbol: Dhcr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022389.2 | NP_071784.1 | 3 - 4 | 531 | 84 | |
XM_006230892.2 | XP_006230954.1 | 2 - 3 | 279 | 84 | ||
XM_006230893.2 | XP_006230955.1 | 2 - 3 | 158 | 84 | ||
XM_006230894.2 | XP_006230956.1 | 3 - 4 | 178 | 84 | ||
XM_008760170.1 | XP_008758392.1 | 3 - 4 | 1265 | 84 | ||
XM_008760171.1 | XP_008758393.1 | 2 - 3 | 926 | 84 | ||
XM_008760172.1 | XP_008758394.1 | 3 - 4 | 825 | 84 | ||
XM_008760173.1 | XP_008758395.1 | 2 - 3 | 805 | 84 | ||
GenBank mRNA | AB016800.1 | - | 1 - 2 | 222 | 84 | |
AF071500.1 | - | 2 - 3 | 134 | 84 | ||
AF272393.1 | - | 3 - 4 | 531 | 84 | ||
BC081688.1 | - | 1 - 2 | 222 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
