Assay Details
Gene Symbol: Dhcr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022389.2 | NP_071784.1 | 6 - 7 | 1058 | 148 | |
XM_006230892.2 | XP_006230954.1 | 5 - 6 | 806 | 148 | ||
XM_006230893.2 | XP_006230955.1 | 5 - 6 | 685 | 148 | ||
XM_006230894.2 | XP_006230956.1 | 6 - 7 | 705 | 148 | ||
XM_008760170.1 | XP_008758392.1 | 6 - 7 | 1792 | 148 | ||
XM_008760171.1 | XP_008758393.1 | 5 - 6 | 1453 | 148 | ||
XM_008760172.1 | XP_008758394.1 | 6 - 7 | 1352 | 148 | ||
XM_008760173.1 | XP_008758395.1 | 5 - 6 | 1332 | 148 | ||
GenBank mRNA | AB016800.1 | - | 4 - 5 | 749 | 148 | |
AF071500.1 | - | 5 - 6 | 661 | 148 | ||
AF272393.1 | - | 6 - 7 | 1058 | 148 | ||
BC081688.1 | - | 4 - 5 | 749 | 7109221 | 148 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
