Assay Details
Gene Symbol: Polr2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006230973.1 | XP_006231035.1 | 1 - 2 | 38 | 79 | |
GenBank mRNA | BC060595.1 | - | 1 - 2 | 47 | 79 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC060595.1, XM_006230973.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
