Assay Details
Gene Symbol: Polr2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053948.4 | NP_446400.1 | 2 - 3 | 197 | 104 | |
XM_006230973.1 | XP_006231035.1 | 4 - 5 | 482 | 104 | ||
GenBank mRNA | BC060550.1 | - | 2 - 3 | 153 | 104 | |
BC060595.1 | - | 4 - 5 | 491 | 104 | ||
FQ223906.1 | - | 4 - 5 | 487 | 104 | ||
FQ224086.1 | - | 2 - 3 | 230 | 104 | ||
FQ232354.1 | - | 2 - 3 | 201 | 104 | ||
Z71925.1 | - | 2 - 3 | 198 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
