Assay Details
Gene Symbol: Polr2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053948.4 | NP_446400.1 | 5 - 6 | 480 | 120 | |
XM_006230973.1 | XP_006231035.1 | 7 - 8 | 765 | 120 | ||
GenBank mRNA | BC060550.1 | - | 5 - 6 | 436 | 120 | |
BC060595.1 | - | 7 - 8 | 774 | 120 | ||
FQ223906.1 | - | 7 - 8 | 770 | 120 | ||
FQ224086.1 | - | 5 - 6 | 513 | 120 | ||
FQ230390.1 | - | 5 - 6 | 443 | 120 | ||
FQ232354.1 | - | 5 - 6 | 484 | 120 | ||
Z71925.1 | - | 5 - 6 | 481 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
