Assay Details
Gene Symbol: Yif1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172017.2 | NP_742014.2 | 4 - 5 | 581 | 58 | |
GenBank mRNA | BC158578.1 | - | 4 - 5 | 562 | 58 | |
CV074812.1 | - | 4 - 5 | 561 | 58 | ||
FQ214813.1 | - | 4 - 5 | 684 | 58 | ||
FQ219390.1 | - | 4 - 5 | 576 | 58 | ||
FQ220202.1 | - | 4 - 5 | 578 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
