Assay Details
Gene Symbol: Yif1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172017.2 | NP_742014.2 | 5 - 6 | 642 | 59 | |
GenBank mRNA | BC158578.1 | - | 5 - 6 | 623 | 59 | |
BG377998.1 | - | 3 - 4 | 59 | |||
CV074812.1 | - | 5 - 6 | 622 | 59 | ||
FQ214813.1 | - | 5 - 6 | 745 | 59 | ||
FQ219390.1 | - | 5 - 6 | 637 | 59 | ||
FQ220202.1 | - | 5 - 6 | 639 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
