Assay Details
Gene Symbol: Yif1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172017.2 | NP_742014.2 | 6 - 7 | 800 | 64 | |
GenBank mRNA | BC158578.1 | - | 6 - 7 | 781 | 64 | |
BG377998.1 | - | 2 - 3 | 64 | |||
FQ214813.1 | - | 6 - 7 | 903 | 64 | ||
FQ219390.1 | - | 6 - 7 | 795 | 64 | ||
FQ220202.1 | - | 6 - 7 | 797 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
