Assay Details
Gene Symbol: Yif1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172017.2 | NP_742014.2 | 7 - 8 | 892 | 68 | |
GenBank mRNA | BC158578.1 | - | 7 - 8 | 873 | 7387164 | 68 |
BG377998.1 | - | 1 - 2 | 68 | |||
FQ219390.1 | - | 7 - 8 | 887 | 68 | ||
FQ220202.1 | - | 7 - 8 | 889 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
