Assay Details
Gene Symbol: Lipa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012732.3 | NP_036864.3 | 2 - 3 | 389 | 105 | |
XM_006231275.2 | XP_006231337.1 | 2 - 3 | 356 | 105 | ||
XM_008760321.1 | XP_008758543.1 | 1 - 2 | 252 | 105 | ||
GenBank mRNA | BC072532.1 | - | 2 - 3 | 389 | 7099460 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
