Assay Details
Gene Symbol: Lipa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012732.3 | NP_036864.3 | 7 - 8 | 1099 | 90 | |
XM_006231275.2 | XP_006231337.1 | 7 - 8 | 1066 | 90 | ||
XM_008760321.1 | XP_008758543.1 | 6 - 7 | 962 | 90 | ||
GenBank mRNA | BC072532.1 | - | 7 - 8 | 1099 | 90 | |
S81497.1 | - | 7 - 8 | 1045 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
