Assay Details
Gene Symbol: Dpp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053748.1 | NP_446200.1 | 16 - 17 | 2054 | 64 | |
XM_006230633.2 | XP_006230695.1 | 17 - 18 | 2241 | 64 | ||
GenBank mRNA | BC107673.1 | - | 17 - 18 | 2065 | 64 | |
D89340.2 | - | 16 - 17 | 2054 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
