Assay Details
Gene Symbol: Oat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022521.3 | NP_071966.1 | 6 - 7 | 875 | 104 | |
GenBank mRNA | BC061551.1 | - | 6 - 7 | 878 | 104 | |
M11842.1 | - | 5 - 6 | 818 | 104 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
