Assay Details
Gene Symbol: Taf6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107575.1 | NP_001101045.1 | 7 - 8 | 696 | 128 | |
XM_006230979.2 | XP_006231041.1 | 7 - 8 | 915 | 128 | ||
XM_006230981.2 | XP_006231043.1 | 7 - 8 | 858 | 128 | ||
XM_006230982.2 | XP_006231044.1 | 8 - 9 | 785 | 128 | ||
XM_006230983.2 | XP_006231045.1 | 7 - 8 | 696 | 128 | ||
XM_008760224.1 | XP_008758446.1 | 7 - 8 | 1167 | 128 | ||
GenBank mRNA | BC168911.1 | - | 7 - 8 | 811 | 128 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
