Assay Details
Gene Symbol: Fgf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257592.1 | XP_006257654.1 | 1 - 2 | 421 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
