Assay Details
Gene Symbol: Nt5c3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107862.1 | NP_001101332.1 | 1 - 2 | 153 | 110 | |
GenBank mRNA | BC166444.1 | - | 1 - 2 | 175 | 110 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC166444.1, NM_001107862.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
