Assay Details
Gene Symbol: Nt5c3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107862.1 | NP_001101332.1 | 4 - 5 | 378 | 125 | |
XM_006236550.2 | XP_006236612.1 | 4 - 5 | 1526 | 125 | ||
XM_006236552.2 | XP_006236614.1 | 3 - 4 | 420 | 125 | ||
GenBank mRNA | BC166444.1 | - | 4 - 5 | 400 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
