Assay Details
Gene Symbol: Nt5c3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107862.1 | NP_001101332.1 | 5 - 6 | 430 | 157 | |
XM_006236550.2 | XP_006236612.1 | 5 - 6 | 1578 | 157 | ||
XM_006236552.2 | XP_006236614.1 | 4 - 5 | 472 | 157 | ||
GenBank mRNA | BC166444.1 | - | 5 - 6 | 452 | 157 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
