Assay Details
Gene Symbol: Nt5c3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107862.1 | NP_001101332.1 | 7 - 8 | 599 | 120 | |
XM_006236550.2 | XP_006236612.1 | 7 - 8 | 1747 | 120 | ||
XM_006236552.2 | XP_006236614.1 | 6 - 7 | 641 | 120 | ||
GenBank mRNA | BC166444.1 | - | 7 - 8 | 621 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
