Assay Details
Gene Symbol: Creb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134621.1 | NP_001128093.1 | 1510 | 73 | ||
XM_008762908.1 | XP_008761130.1 | 5 - 6 | 1265 | 73 | ||
XM_008762909.1 | XP_008761131.1 | 9 - 10 | 1660 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
