Assay Details
Gene Symbol: Trim46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107691.1 | NP_001101161.1 | 2 - 3 | 351 | 112 | |
XM_008761170.1 | XP_008759392.1 | 2 - 3 | 468 | 112 | ||
XM_008761171.1 | XP_008759393.1 | 2 - 3 | 315 | 112 | ||
XM_008761172.1 | XP_008759394.1 | 2 - 3 | 314 | 112 | ||
XM_008761173.1 | XP_008759395.1 | 2 - 3 | 315 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
