Assay Details
Gene Symbol: Clk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014254.1 | NP_001014276.1 | 5 - 6 | 823 | 88 | |
XM_008761185.1 | XP_008759407.1 | 5 - 6 | 1028 | 88 | ||
XM_008761186.1 | XP_008759408.1 | 6 - 7 | 895 | 88 | ||
XM_008761187.1 | XP_008759409.1 | 5 - 6 | 1025 | 88 | ||
XM_008761188.1 | XP_008759410.1 | 5 - 6 | 1007 | 88 | ||
XM_008761189.1 | XP_008759411.1 | 5 - 6 | 1004 | 88 | ||
XM_008761190.1 | XP_008759412.1 | 5 - 6 | 1004 | 88 | ||
XM_008761191.1 | XP_008759413.1 | 5 - 6 | 1001 | 88 | ||
XM_008761192.1 | XP_008759414.1 | 5 - 6 | 874 | 88 | ||
XM_008761193.1 | XP_008759415.1 | 2 - 3 | 675 | 88 | ||
XM_008761194.1 | XP_008759416.1 | 2 - 3 | 154 | 88 | ||
GenBank mRNA | BC083788.1 | - | 5 - 6 | 823 | 7098251 | 88 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the XM_008761194.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
