Assay Details
Gene Symbol: Clk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014254.1 | NP_001014276.1 | 7 - 8 | 1102 | 92 | |
XM_008761185.1 | XP_008759407.1 | 7 - 8 | 1307 | 92 | ||
XM_008761186.1 | XP_008759408.1 | 8 - 9 | 1174 | 92 | ||
XM_008761187.1 | XP_008759409.1 | 7 - 8 | 1304 | 92 | ||
XM_008761188.1 | XP_008759410.1 | 7 - 8 | 1286 | 92 | ||
XM_008761189.1 | XP_008759411.1 | 7 - 8 | 1283 | 92 | ||
XM_008761190.1 | XP_008759412.1 | 7 - 8 | 1283 | 92 | ||
XM_008761191.1 | XP_008759413.1 | 7 - 8 | 1280 | 92 | ||
XM_008761192.1 | XP_008759414.1 | 7 - 8 | 1153 | 92 | ||
XM_008761193.1 | XP_008759415.1 | 4 - 5 | 954 | 92 | ||
XM_008761194.1 | XP_008759416.1 | 4 - 5 | 433 | 92 | ||
GenBank mRNA | BC083788.1 | - | 7 - 8 | 1102 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
