Assay Details
Gene Symbol: Clk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014254.1 | NP_001014276.1 | 9 - 10 | 1325 | 130 | |
XM_008761185.1 | XP_008759407.1 | 9 - 10 | 1530 | 130 | ||
XM_008761186.1 | XP_008759408.1 | 10 - 11 | 1397 | 130 | ||
XM_008761187.1 | XP_008759409.1 | 9 - 10 | 1527 | 130 | ||
XM_008761188.1 | XP_008759410.1 | 9 - 10 | 1509 | 130 | ||
XM_008761189.1 | XP_008759411.1 | 9 - 10 | 1506 | 130 | ||
XM_008761190.1 | XP_008759412.1 | 9 - 10 | 1506 | 130 | ||
XM_008761191.1 | XP_008759413.1 | 9 - 10 | 1503 | 130 | ||
XM_008761192.1 | XP_008759414.1 | 9 - 10 | 1376 | 130 | ||
XM_008761193.1 | XP_008759415.1 | 6 - 7 | 1177 | 130 | ||
XM_008761194.1 | XP_008759416.1 | 6 - 7 | 656 | 130 | ||
GenBank mRNA | BC083788.1 | - | 9 - 10 | 1325 | 130 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
