Assay Details
Gene Symbol: Mrpl30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766999.1 | XP_008765221.1 | 1 - 2 | 91 | 59 | |
GenBank mRNA | BC168851.1 | - | 1 - 2 | 27 | 59 |
Gene Symbol: NEWGENE_1308196 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008767056.1 | XP_008765278.1 | 129 | 59 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC168851.1, XM_008766999.1, XM_008767056.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
