Assay Details
Gene Symbol: Eef2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017245.2 | NP_058941.1 | 10 - 11 | 2451 | 63 | |
GenBank mRNA | AF000576.1 | - | 2 - 3 | 743 | 63 | |
BC066661.1 | - | 10 - 11 | 2451 | 5623322 | 63 | |
FQ220112.1 | - | 2 - 3 | 687 | 63 | ||
FQ229435.1 | - | 2 - 3 | 505 | 63 | ||
K03502.1 | - | 3 - 4 | 845 | 63 | ||
Y07504.1 | - | 10 - 11 | 2437 | 63 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
