Assay Details
Gene Symbol: Gadd45b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008321.1 | NP_001008322.1 | 2 - 3 | 383 | 56 | |
XM_006240888.2 | XP_006240950.1 | 2 - 3 | 392 | 56 | ||
GenBank mRNA | BC085337.1 | - | 2 - 3 | 383 | 7302492 | 56 |
FQ235047.1 | - | 2 - 3 | 392 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
