Assay Details
Gene Symbol: Csnk1g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033870.1 | NP_001029042.1 | 7 - 8 | 1226 | 122 | |
NM_023102.2 | NP_075590.2 | 7 - 8 | 1223 | 122 | ||
XM_006241000.2 | XP_006241062.1 | 6 - 7 | 2601 | 122 | ||
XM_006241001.2 | XP_006241063.1 | 6 - 7 | 1309 | 122 | ||
XM_006241002.2 | XP_006241064.1 | 7 - 8 | 2521 | 122 | ||
XM_006241003.2 | XP_006241065.1 | 6 - 7 | 1951 | 122 | ||
XM_008765130.1 | XP_008763352.1 | 6 - 7 | 1309 | 122 | ||
GenBank mRNA | BC072533.1 | - | 7 - 8 | 1226 | 7098992 | 122 |
BC099758.1 | - | 1 - 2 | 85 | 122 | ||
U22297.1 | - | 6 - 7 | 957 | 122 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
