Assay Details
Gene Symbol: Csnk1g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023102.2 | NP_075590.2 | 12 - 13 | 1707 | 85 | |
XM_006241001.2 | XP_006241063.1 | 11 - 12 | 1793 | 85 | ||
GenBank mRNA | U22297.1 | - | 11 - 12 | 1441 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
