Assay Details
Gene Symbol: Dapk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022546.1 | NP_071991.1 | 4 - 5 | 666 | 73 | |
XM_006240991.1 | XP_006241053.1 | 4 - 5 | 797 | 73 | ||
XM_006240993.2 | XP_006241055.1 | 4 - 5 | 904 | 73 | ||
XM_008765129.1 | XP_008763351.1 | 4 - 5 | 1130 | 73 | ||
GenBank mRNA | AJ006971.1 | - | 4 - 5 | 666 | 73 | |
BC062076.1 | - | 4 - 5 | 667 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
