Assay Details
Gene Symbol: Slc1a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032065.1 | NP_114454.1 | 6 - 7 | 1313 | 75 | |
XM_008765182.1 | XP_008763404.1 | 6 - 7 | 1487 | 75 | ||
GenBank mRNA | AY286330.1 | - | 6 - 7 | 1307 | 75 | |
BC098912.1 | - | 6 - 7 | 1337 | 7315936 | 75 | |
U89608.1 | - | 6 - 7 | 1313 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
