Assay Details
Gene Symbol: Madcam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019317.1 | NP_062190.1 | 2 - 3 | 334 | 133 | |
XM_008765179.1 | XP_008763401.1 | 2 - 3 | 646 | 133 | ||
GenBank mRNA | D87840.1 | - | 2 - 3 | 334 | 133 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
