Assay Details
Gene Symbol: Gna11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031033.1 | NP_112295.1 | 6 - 7 | 887 | 112 | |
GenBank mRNA | AF239674.1 | - | 6 - 7 | 887 | 112 | |
L37293.1 | - | 1 - 2 | 158 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
