Assay Details
Gene Symbol: Sgta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022703.3 | NP_073194.1 | 3 - 4 | 269 | 143 | |
XM_006240997.1 | XP_006241059.1 | 3 - 4 | 371 | 143 | ||
GenBank mRNA | AF368278.1 | - | 3 - 4 | 281 | 143 | |
AJ222724.1 | - | 3 - 4 | 257 | 143 | ||
BC087642.1 | - | 3 - 4 | 269 | 7308544 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
