Assay Details
Gene Symbol: Sgta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022703.3 | NP_073194.1 | 7 - 8 | 700 | 73 | |
XM_006240996.1 | XP_006241058.1 | 7 - 8 | 805 | 73 | ||
XM_006240997.1 | XP_006241059.1 | 7 - 8 | 802 | 73 | ||
XM_006240998.2 | XP_006241060.1 | 7 - 8 | 2645 | 73 | ||
GenBank mRNA | AF368278.1 | - | 7 - 8 | 712 | 73 | |
AJ222724.1 | - | 7 - 8 | 688 | 73 | ||
BC087642.1 | - | 7 - 8 | 700 | 7308544 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
