Assay Details
Gene Symbol: Pigl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138901.2 | NP_620256.1 | 4 - 5 | 501 | 93 | |
XM_008767767.1 | XP_008765989.1 | 4 - 5 | 497 | 93 | ||
GenBank mRNA | BC074020.1 | - | 4 - 5 | 501 | 7104008 | 93 |
D88364.1 | - | 4 - 5 | 882 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
